Anti-CSF1 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB9693-100EA 4365.25 DKK
ABCAAB9693-100
Anti-CSF1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CSF1 (colony stimulating factor 1 (macrophage))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human