Anti-CCL7 Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9681-100EA 5330 DKK
ABCAAB9681-100
Anti-CCL7 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CCL7 (chemokine (C-C motif) ligand 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR