Anti-Interferon gamma Mouse Monoclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9658-500EA 5330 DKK
ABCAAB9658-500
Anti-Interferon gamma Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Interferon gamma
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR