Anti-IL3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9620-100EA 5330 DKK
ABCAAB9620-100
Anti-IL3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IL3 (interleukin 3 (colony-stimulating factor, multiple))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR