Anti-INS Mouse Monoclonal Antibody [clone: E2E3]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9569-100EA 5910 DKK
ABCAAB9569-100
Anti-INS Mouse Monoclonal Antibody [clone: E2E3]
Antistoffer
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone: E2E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR