Anti-FLT1 Mouse Monoclonal Antibody [clone: Flt-1/EWC]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9540-100EA 5610 DKK
ABCAAB9540-100
Anti-FLT1 Mouse Monoclonal Antibody [clone: Flt-1/EWC]
Antistoffer
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: monoclonal
Clone: Flt-1/EWC
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Dog, Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR