Anti-GLB1 Chicken Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9361-250EA 6250 DKK
ABCAAB9361-250
Anti-GLB1 Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GLB1 (galactosidase, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR