Anti-Phosphothreonine Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9338-50EA 5760 DKK
ABCAAB9338-50
Anti-Phosphothreonine Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: Phosphothreonine
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR