Anti-NTF4 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9330-500EA 4300 DKK
ABCAAB9330-500
Anti-NTF4 Mouse Monoclonal Antibody
Type: Primary
Antigen: NTF4 (neurotrophin 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR