Anti-COL7 Mouse Monoclonal Antibody [clone: LH7.2]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9276-100EA 4060 DKK
ABCAAB9276-100
Anti-COL7 Mouse Monoclonal Antibody [clone: LH7.2]
Type: Primary
Antigen: COL7 (collagen, type VII)
Clonality: monoclonal
Clone: LH7.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR