Anti-MMP13 Mouse Monoclonal Antibody [clone: M53]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9127-50EA 4140 DKK
ABCAAB9127-50
Anti-MMP13 Mouse Monoclonal Antibody [clone: M53]
Type: Primary
Antigen: MMP13 (matrix metallopeptidase 13 (collagenase 3))
Clonality: monoclonal
Clone: M53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR