Anti-Myc Tag Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9106-100EA 4806.36 DKK
ABCAAB9106-100
Anti-Myc Tag Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Myc Tag (Myc tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR