Anti-HIST4H4 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB9051-100EA 4402.56 DKK
ABCAAB9051-100
Anti-HIST4H4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu