Anti-UMOD Sheep Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9029-1EA 5330 DKK
ABCAAB9029-1
Anti-UMOD Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: UMOD (uromodulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR