Anti-NCAM1 Mouse Monoclonal Antibody [clone: RNL-1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB9018-100EA 5990 DKK
ABCAAB9018-100
Anti-NCAM1 Mouse Monoclonal Antibody [clone: RNL-1]
Antistoffer
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: RNL-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR