Anti-ITGA6A Mouse Monoclonal Antibody [clone: 1A10]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8989-100EA 4060 DKK
ABCAAB8989-100
Anti-ITGA6A Mouse Monoclonal Antibody [clone: 1A10]
Type: Primary
Antigen: ITGA6A (integrin, alpha 6A)
Clonality: monoclonal
Clone: 1A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR