Anti-ITGA3A Mouse Monoclonal Antibody [clone: 29A3]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8985-100EA 5370 DKK
ABCAAB8985-100
Anti-ITGA3A Mouse Monoclonal Antibody [clone: 29A3]
Antistoffer
Type: Primary
Antigen: ITGA3A (integrin, alpha 3A)
Clonality: monoclonal
Clone: 29A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR