Anti-LMNA Mouse Monoclonal Antibody [clone: 133A2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8980-100EA 6040 DKK
ABCAAB8980-100
Anti-LMNA Mouse Monoclonal Antibody [clone: 133A2]
Antistoffer
Type: Primary
Antigen: LMNA (lamin A/C)
Clonality: monoclonal
Clone: 133A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR