Anti-DES Mouse Monoclonal Antibody [clone: RD301]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8976-100EA 5790 DKK
ABCAAB8976-100
Anti-DES Mouse Monoclonal Antibody [clone: RD301]
Antistoffer
Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone: RD301
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR