Anti-Mitotic Cells Mouse Monoclonal Antibody [clone: 8B3G]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8956-100EA 5500 DKK
ABCAAB8956-100
Anti-Mitotic Cells Mouse Monoclonal Antibody [clone: 8B3G]
Antistoffer
Type: Primary
Antigen: Mitotic Cells
Clonality: monoclonal
Clone: 8B3G
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR