Anti-Salivary Amylase Sheep Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8944-1EA 4060 DKK
ABCAAB8944-1
Anti-Salivary Amylase Sheep Polyclonal Antibody
Type: Primary
Antigen: Salivary Amylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR