Anti-L Glutamate Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8893-50EA 4300 DKK
ABCAAB8893-50
Anti-L Glutamate Mouse Monoclonal Antibody
Type: Primary
Antigen: L Glutamate
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR