Anti-GABA Rabbit Polyclonal Antibody (Unconjugated)

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8891-50EA 5550 DKK
ABCAAB8891-50
Anti-GABA Rabbit Polyclonal Antibody (Unconjugated)
Antistoffer
Type: Primary
Antigen: GABA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR