Anti-CD99 Mouse Monoclonal Antibody [clone: 12E7]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8855-250EA 5750 DKK
ABCAAB8855-250
Anti-CD99 Mouse Monoclonal Antibody [clone: 12E7]
Antistoffer
Type: Primary
Antigen: CD99 (CD99 molecule)
Clonality: monoclonal
Clone: 120000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR