Anti-IgA/IgG/IgM Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8525-1EA 2280 DKK
ABCAAB8525-1
Anti-IgA/IgG/IgM Rabbit Polyclonal Antibody
Antistoffer
Type: Secondary
Antigen: Sheep IgG+IgM+IgA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR