Anti-Lipid A Mouse Monoclonal Antibody [clone: 26-5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8467-1EA 5630 DKK
ABCAAB8467-1
Anti-Lipid A Mouse Monoclonal Antibody [clone: 26-5]
Antistoffer
Type: Primary
Antigen: Lipid A
Clonality: monoclonal
Clone: 26-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR