Anti-CGB5 Mouse Monoclonal Antibody [clone: 2092]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8466-1EA 5630 DKK
ABCAAB8466-1
Anti-CGB5 Mouse Monoclonal Antibody [clone: 2092]
Antistoffer
Type: Primary
Antigen: CGB5 (chorionic gonadotropin, beta polypeptide 5)
Clonality: monoclonal
Clone: 2092
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR