Anti-IL1B Mouse Monoclonal Antibody [clone: 11E5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8320-100EA 5500 DKK
ABCAAB8320-100
Anti-IL1B Mouse Monoclonal Antibody [clone: 11E5]
Antistoffer
Type: Primary
Antigen: IL1B (interleukin 1, beta)
Clonality: monoclonal
Clone: 1100000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR