Anti-Insulin + Proinsulin Mouse Monoclonal Antibody [clone: D3E7]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8305-250EA 4990 DKK
ABCAAB8305-250
Anti-Insulin + Proinsulin Mouse Monoclonal Antibody [clone: D3E7]
Type: Primary
Antigen: Insulin + Proinsulin
Clonality: monoclonal
Clone: D3E7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR