Anti-Proinsulin Mouse Monoclonal Antibody [clone: 3A1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8301-100EA 5620 DKK
ABCAAB8301-100
Anti-Proinsulin Mouse Monoclonal Antibody [clone: 3A1]
Antistoffer
Type: Primary
Antigen: Proinsulin
Clonality: monoclonal
Clone: 3A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR