Anti-TNNI3 Mouse Monoclonal Antibody [clone: 5E6]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8291-200EA 4140 DKK
ABCAAB8291-200
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 5E6]
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: monoclonal
Clone: 5000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR