Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-148]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8220-100EA 5810 DKK
ABCAAB8220-100
Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-148]
Antistoffer
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone: MEM-148
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR