Anti-Lambda Light chain Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8178-500EA 4300 DKK
ABCAAB8178-500
Anti-Lambda Light chain Rabbit Polyclonal Antibody
Type: Primary
Antigen: Lambda Light chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR