Anti-DIABLO Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB8115-100EA 4098.45 DKK
ABCAAB8115-100
Anti-DIABLO Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DIABLO (diablo, IAP-binding mitochondrial protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat