Anti-NFkB Inducing Kinase NIK Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8111-100EA 5330 DKK
ABCAAB8111-100
Anti-NFkB Inducing Kinase NIK Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NFkB Inducing Kinase NIK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR