Anti-TNF Rat Monoclonal Antibody [clone: V1q(H8)]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8097-50EA 5670 DKK
ABCAAB8097-50
Anti-TNF Rat Monoclonal Antibody [clone: V1q(H8)]
Antistoffer
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: V1q(H8)
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR