Anti-Endothelium Mouse Monoclonal Antibody [clone: EN4]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8087-1EA 5810 DKK
ABCAAB8087-1
Anti-Endothelium Mouse Monoclonal Antibody [clone: EN4]
Antistoffer
Type: Primary
Antigen: Endothelium
Clonality: monoclonal
Clone: EN4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR