Anti-HLA DQ Mouse Monoclonal Antibody [clone: BL-A/G3]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB8083-1EA 4300 DKK
ABCAAB8083-1
Anti-HLA DQ Mouse Monoclonal Antibody [clone: BL-A/G3]
Type: Primary
Antigen: HLA DQ
Clonality: monoclonal
Clone: BL-A/G3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR