Anti-Tropomyosin Mouse Monoclonal Antibody [clone: CH1]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB7786-250EA 4590 DKK
ABCAAB7786-250
Anti-Tropomyosin Mouse Monoclonal Antibody [clone: CH1]
Type: Primary
Antigen: Tropomyosin
Clonality: monoclonal
Clone: CH1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR