Anti-COL3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB7778-100EA 6250 DKK
ABCAAB7778-100
Anti-COL3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR