Anti-Interferon gamma Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB7740-500EA 5630 DKK
ABCAAB7740-500
Anti-Interferon gamma Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Interferon gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR