Anti-INA Mouse Monoclonal Antibody [clone: 2E3]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB7654-250EA 5880 DKK
ABCAAB7654-250
Anti-INA Mouse Monoclonal Antibody [clone: 2E3]
Antistoffer
Type: Primary
Antigen: INA (internexin neuronal intermediate filament protein, alpha)
Clonality: monoclonal
Clone: 2000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR