Anti-SIRT7 Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB87947-50EA 0 DKK
BIRBORB87947-50
Anti-SIRT7 Rabbit Polyclonal Antibody
Antistoffer
  • Immunochemistry: 5 μg/ml; Western Blot: 1-3 μg/ml
Type: Primary
Antigen: SIRT7 (sirtuin 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human