Anti-SIRT5 Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB87946-50EA 0 DKK
BIRBORB87946-50
Anti-SIRT5 Rabbit Polyclonal Antibody
Antistoffer
Immunochemistry: 5 ?g/ml; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: SIRT5 (sirtuin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat