Anti-Psoriasin Mouse Monoclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB87943-50EA 0 DKK
BIRBORB87943-50
Anti-Psoriasin Mouse Monoclonal Antibody
Antistoffer
Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: Psoriasin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human