Anti-MBD3 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB87939-50EA 0 DKK
BIRBORB87939-50
Anti-MBD3 Mouse Monoclonal Antibody
Antistoffer
Immunochemistry: 10 ?g/ml; Western Blot: 2 ?g/ml

Type: Primary
Antigen: MBD3 (methyl-CpG binding domain protein 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR