Anti-MAPK9 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB87938-50EA 0 DKK
BIRBORB87938-50
Anti-MAPK9 Rabbit Polyclonal Antibody
Antistoffer
Immunochemistry: 10 ?g/ml; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: MAPK9 (mitogen-activated protein kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR