Anti-HMGA1 Rabbit Polyclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB87923-50EA 0 DKK
BIRBORB87923-50
Anti-HMGA1 Rabbit Polyclonal Antibody
Antistoffer
Immunochemistry: 10 ?g/ml; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: HMGA1 (high mobility group AT-hook 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu