Anti-HDAC3 Rabbit polyclonal antibody unconjugated

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB87918-50EA 0 DKK
BIRBORB87918-50
Anti-HDAC3 Rabbit polyclonal antibody unconjugated
Antistoffer
Immunochemistry: 10 ?g/ml: 2 ?g/ml

Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR