Anti-GOLM1 Rabbit polyclonal antibody unconjugated
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB87916-50EA 0 DKK
BIRBORB87916-50
Anti-GOLM1 Rabbit polyclonal antibody unconjugated
Antistoffer
ELISA: 1:100-1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: GOLM1 (golgi membrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human